Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR2C3 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB048563 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB048563 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB048563 Supplier Novus Biologicals Supplier No. NBP297856UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR2C3 Polyclonal specifically detects OR2C3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen OR2C3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias olfactory receptor 2C3, Olfactory receptor 2C4, Olfactory receptor 2C5, Olfactory receptor OR1-30, olfactory receptor, family 2, subfamily C, member 3, olfactory receptor, family 2, subfamily C, member 4, olfactory receptor, family 2, subfamily C, member 5 pseudogene, OR2C4, OR2C5P, OST742
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human OR2C3 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81472
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.