Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR5B3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309798100UL
Description
OR5B3 Polyclonal specifically detects OR5B3 in Human samples. It is validated for Western Blot.Specifications
OR5B3 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Olfactory receptor 5B13, olfactory receptor 5B3, Olfactory receptor OR11-239, olfactory receptor, family 5, subfamily B, member 13, olfactory receptor, family 5, subfamily B, member 3, OR11-239, OR5B13, OST129 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR5B3 (NP_001005469). Peptide sequence SSSHSMDTDKMAPVFYTMVIPMLNPLVYSLRNKEVKSAFKKVVEKAKLSV | |
100 μg | |
Signal Transduction | |
441608 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction