Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR5F1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB329515 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329515 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329515 Supplier Novus Biologicals Supplier No. NBP297876F

Rabbit Polyclonal Antibody

OR5F1 Polyclonal antibody specifically detects OR5F1 in Human samples. It is validated for Western Blot

Specifications

Antigen OR5F1
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias olfactory receptor 5F1, Olfactory receptor OR11-167, olfactory receptor, family 5, subfamily F, member 1, OR11-10Olfactory receptor 11-10
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human OR5F1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338674
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.