Learn More
Description
Specifications
Specifications
| Antigen | OR6B3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | olfactory receptor 6B3, Olfactory receptor OR2-2, olfactory receptor, family 6, subfamily B, member 3, olfactory receptor, family 6, subfamily B, member 3 pseudogene, OR6B3P, OR6B3Q |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR6B3 (NP_775486). Peptide sequence RIPSATGCWRAFFTCASHLTVVTVFYTALLFMYVRPQAIDSRSSNKLISV |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
