Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR6X1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB293131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293131 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293131 Supplier Novus Biologicals Supplier No. NBP326089G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR6X1 Polyclonal antibody specifically detects OR6X1 in Human samples. It is validated for Western Blot

Specifications

Antigen OR6X1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias olfactory receptor 6X1, olfactory receptor, family 6, subfamily X, member 1, OR11-270
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human OR6X1
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390260
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.