Learn More
ORAI1 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 100401P17
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-ORAI-1 antibody is directed against the human ORAI-1. Cross-reactivity with ORAI1 from other sources have not been determined.
ORAI1 is a plasma membrane protein essential for store-operated calcium entry.Specifications
| ORAI1 | |
| Polyclonal | |
| Unconjugated | |
| Orai1 | |
| calcium release-activated calcium channel 1; Calcium release-activated calcium channel protein 1; calcium release-activated calcium modulator 1; Cracm1; D730049H07Rik; FLJ14466; IMD9; ORAI calcium release-activated calcium modulator 1; ORAI1; orai-1; ORAT1; Protein orai-1; RGD1311873; TAM2; TMEM142A; Transmembrane protein 142A | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
| Immunoprecipitation, Western Blot | |
| 45 mg/mL | |
| 0.01M sodium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| Q96D31 | |
| Orai1 | |
| This whole rabbit serum was prepared by repeated immunizations with a recombinant fusion protein. | |
| RUO | |
| 84876 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.