Learn More
ORAI1 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $661.74
Specifications
| Antigen | ORAI1 |
|---|---|
| Concentration | 45 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0156
![]()
|
Rockland Immunochemicals
100401P17S |
25 μL |
Each for $178.35
|
|
|||||
50-199-0155
![]()
|
Rockland Immunochemicals
100401P17 |
100 μL |
Each for $661.74
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-ORAI-1 antibody is directed against the human ORAI-1. Cross-reactivity with ORAI1 from other sources have not been determined.
ORAI1 is a plasma membrane protein essential for store-operated calcium entry.Specifications
| ORAI1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96D31 | |
| 84876 | |
| This whole rabbit serum was prepared by repeated immunizations with a recombinant fusion protein. | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Orai1 |
| 45 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.01M sodium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| calcium release-activated calcium channel 1; Calcium release-activated calcium channel protein 1; calcium release-activated calcium modulator 1; Cracm1; D730049H07Rik; FLJ14466; IMD9; ORAI calcium release-activated calcium modulator 1; ORAI1; orai-1; ORAT1; Protein orai-1; RGD1311873; TAM2; TMEM142A; Transmembrane protein 142A | |
| Orai1 | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.