Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orai2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB216804 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB216804 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB216804 Supplier Novus Biologicals Supplier No. NBP177283AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Orai2 Polyclonal antibody specifically detects Orai2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Orai2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias CAP-binding protein complex interacting protein 2, CAP-binding protein complex-interacting protein 2, CBCIP2C7orf19FLJ12474, chromosome 7 open reading frame 19, FLJ14733, H_NH0514P08.8, MEM142B, ORAI calcium release-activated calcium modulator 2, protein orai-2, putative protein ORAI2-2, TMEM142B, Transmembrane protein 142BFLJ44818
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human ORAI2. The immunogen is located within the last 50 amino acids of ORAI2. Amino Acid Squence: HKTERHNREIEELHK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 80228
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.