Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orai2 Antibody (OTI8D11), PE, Novus Biologicals™
SDP

Catalog No. NB302164 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302164 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302164 Supplier Novus Biologicals Supplier No. NBP273148PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Orai2 Monoclonal antibody specifically detects Orai2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Orai2
Applications Western Blot
Classification Monoclonal
Clone OTI8D11
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias CAP-binding protein complex interacting protein 2, CAP-binding protein complex-interacting protein 2, CBCIP2C7orf19FLJ12474, chromosome 7 open reading frame 19, FLJ14733, H_NH0514P08.8, MEM142B, ORAI calcium release-activated calcium modulator 2, protein orai-2, putative protein ORAI2-2, TMEM142B, Transmembrane protein 142BFLJ44818
Host Species Mouse
Immunogen Full length human recombinant protein of human ORAI2 (NP_116220) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 80228
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.