Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORAI3 Antibody (1B4F1), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB203089 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB203089 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB203089 Supplier Novus Biologicals Supplier No. NBP241327UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ORAI3 Monoclonal antibody specifically detects ORAI3 in Human, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen ORAI3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1B4F1
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias ORAI calcium release-activated calcium modulator 3, TMEM142C
Host Species Mouse
Immunogen A 19 amino acid synthetic peptide from near the carboxy terminus of human ORAI3. Amino Acid Squence: ASNLPRSSASAAPSQAEPA
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 93129
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.