Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORAI3 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB341493 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341493 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341493 Supplier Novus Biologicals Supplier No. NBP282014MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ORAI3 Polyclonal antibody specifically detects ORAI3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen ORAI3
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ORAI calcium release-activated calcium modulator 3, TMEM142C
Host Species Rabbit
Immunogen ORAI3 antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human ORAI3. The immunogen is located within amino acids 200 - 250 of ORAI3. Amino Acid Squence: ASNLPRSSASAAPSQAEPA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 93129
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.