Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products Orbiter Planetarium
SCI_ED

Catalog No. p-2373651 Shop All American Educational Products LLC Products
Click to view available options
Product Type:
Illuminated Orbiter
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Lesson plan

Explain the cycles of day and night, the seasons, the phases of the moon and many other astronomy concepts.

  • Hand-operated gear-and-chain drive rotates the Earth on its axis, orients the axis in relation to the sun and revolves the Earth around the sun
  • Features a 6 in. sun (15.2cm), a 15 in. arm (38.1cm), a 4 in. geophysical Earth globe (10.2cm), a cratered moon and a transparent hemisphere screen

Specifications

Grade 3 to 12
For Use With (Application) Explains the cycles of day and night, the seasons, the phases of the moon and many other astronomy concepts
Includes Lesson plan, 6 in. sun, 15 in. arm, 4 in. geophysical earth globe, cratered moon and transparent hemisphere screen
Product Type Illuminated Orbiter
Quantity Each
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.