Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORC1 Antibody (7F6/1), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB390612 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390612 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390612 Supplier Novus Biologicals Supplier No. NBP308793AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ORC1 Monoclonal antibody specifically detects ORC1 in Human samples. It is validated for ELISA

Specifications

Antigen ORC1
Applications ELISA
Classification Monoclonal
Clone 7F6/1
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias HS, HSORC1, ORC1Lorigin recognition complex subunit 1, origin recognition complex, subunit 1, origin recognition complex, subunit 1 (yeast homolog)-like, origin recognition complex, subunit 1 homolog, origin recognition complex, subunit 1 homolog (S. cerevisiae), origin recognition complex, subunit 1-like, origin recognition complex, subunit 1-like (yeast), PARC1origin recognition complex 1, Replication control protein 1
Host Species Mouse
Immunogen Recombinant full-length human ORC1 protein (Uniprot: Q13415)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4998
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.