Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORC3L Antibody, Novus Biologicals™
SDP

Catalog No. NB100796 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100796 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100796 Supplier Novus Biologicals Supplier No. NB100796
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ORC3L Polyclonal specifically detects ORC3L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen ORC3L
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 μg/mL, Immunohistochemistry 3.75 μg/mL, Immunohistochemistry-Paraffin 3.75 μg/mL, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_036513.
Gene Alias IMAGE50150, LAT, LATHEOorigin recognition complex, subunit 3-like (yeast), ORC3Lhomolog of latheo, Drosophila, origin recognition complex subunit 3, Origin recognition complex subunit Latheo, origin recognition complex, subunit 3, origin recognition complex, subunit 3 (yeast homolog)-like, origin recognition complex, subunit 3 honolog, origin recognition complex, subunit 3 honolog (yeast), origin recognition complex, subunit 3-like
Gene Symbols ORC3
Host Species Goat
Immunogen Peptide with sequence QKTDHVARLTWGGC corresponding to C-Terminus according to NP_862820.1, NP_036513.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23595
Test Specificity This antibody is expected to recognize both reported isoforms (NP_862820.1 and NP_036513.2).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.