Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orencia (Abatacept) Antibody (309C2.01), Biotin, Novus Biologicals™
SDP

Catalog No. DDX540B1001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX540B1001 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX540B1001 Supplier Novus Biologicals Supplier No. DDX5040B1000.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Orencia (Abatacept) Monoclonal specifically detects Orencia (Abatacept) in Human samples. It is validated for ELISA.

Specifications

Antigen Orencia (Abatacept)
Applications ELISA
Classification Monoclonal
Clone 309C2.01
Concentration 0.5 mg/mL
Conjugate Biotin
Dilution ELISA
Formulation PBS 50% glycerol with No Preservative
Host Species Mouse
Immunogen Abatacept
Purification Method Affinity Purified
Quantity 0.1 mg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.