Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orexin A/Hypocretin-1 Antibody (MM0500-8G22), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB361662 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361662 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361662 Supplier Novus Biologicals Supplier No. NBP211833MFV500

Mouse Monoclonal Antibody

Orexin A/Hypocretin-1 Monoclonal antibody specifically detects Orexin A/Hypocretin-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen Orexin A/Hypocretin-1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MM0500-8G22
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias hypocretin (orexin) neuropeptide precursor, NRCLP1, orexin, OXHcrt, PPORX, PPOXHypocretin
Host Species Mouse
Immunogen Human Orexin A/Hypocretin-1 peptide
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3060
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.