Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orexin B/Hypocretin-2 Antibody (MM0501-5G21), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB211834405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211834405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211834405 Supplier Novus Biologicals Supplier No. NBP211834AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Orexin B/Hypocretin-2 Monoclonal specifically detects Orexin B/Hypocretin-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Orexin B/Hypocretin-2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MM0501-5G21
Conjugate Alexa Fluor 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HCRT, Hcrt 2, Hcrt2, Hypocretin, Hypocretin (orexin) neuropeptide precursor, Hypocretin 2, Hypocretin B, Hypocretin precursor, Hypocretin2, Orexin, Orexin neuropeptide precursor, Orexin precursor, OrexinB, PPORX, PPOX
Gene Symbols HCRT
Host Species Mouse
Immunogen Human Orexin B/Hypocretin-2 peptide
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, GPCR, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3060
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.