Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orexin R2/HCRTR2 Antibody (456738), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB303302 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303302 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303302 Supplier Novus Biologicals Supplier No. FAB5246S

Mouse Monoclonal Antibody

Orexin R2/HCRTR2 Monoclonal antibody specifically detects Orexin R2/HCRTR2 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Orexin R2/HCRTR2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 456738
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias hypocretin (orexin) receptor 2, Hypocretin receptor type 2, orexin receptor type 2, Ox-2-R, Ox2-R, OX2R
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Orexin R2/HCRTR2, Met1-Trp444, Accession # AAC39602
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, GPCR, Immunology, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3062
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.