Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Orexin Receptor 2 Monoclonal Antibody (456738)
GREENER_CHOICE

Catalog No. PIMA524178
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524178 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524178 Supplier Invitrogen™ Supplier No. MA524178
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody stains human Orexin R2/HCRTR2 transfectants but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.

The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein binds the hypothalamic neuropeptides orexin A and orexin B. A related gene encodes a G-protein coupled receptor that selectively binds orexin A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Orexin Receptor 2
Applications Flow Cytometry
Classification Monoclonal
Clone 456738
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene HCRTR2
Gene Accession No. O43614
Gene Alias HCRTR1; Hcrtr2; hypocretin (orexin) receptor 2; hypocretin receptor 2; hypocretin receptor type 2; mOX2aR; mOX2bR; Mox2r; mOXR2; orexin receptor 1; orexin receptor 2; orexin receptor type 2; orexin receptor-1; orexin receptor-2; orexin type-2 receptor; OTTHUMP00000039969; OX1R; ox2r; ox-2-R; ox2-R
Gene Symbols HCRTR2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Orexin R2/HCRTR2 Met1-Trp444.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3062
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.