Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORF73/HHV8 Antibody (HHV8/3633R), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB392347 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392347 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392347 Supplier Novus Biologicals Supplier No. NBP308397AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ORF73/HHV8 Monoclonal antibody specifically detects ORF73/HHV8 in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ORF73/HHV8
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HHV8/3633R
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV
Host Species Rabbit
Immunogen Recombinant protein corresponding to the latent nuclear antigen 1 molecule of ORF73/HHV8
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 935114
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.