Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORF73/HHV8 Rat anti-Virus, Clone: LN53, Novus Biologicals™
SDP

Catalog No. NB029773 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB029773 0.025 mg
NB029774 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB029773 Supplier Novus Biologicals Supplier No. NBP1227670.025MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

ORF73/HHV8 Monoclonal specifically detects ORF73/HHV8 in Virus samples. It is validated for Immunoassay, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen ORF73/HHV8
Applications Immunoassay, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LN53
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay
Formulation PBS with 0.02% Sodium Azide
Gene Alias HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV
Gene Symbols HHV8GP78
Host Species Rat
Immunogen Recombinant protein corresponding to the latent nuclear antigen-1 molecule of HHV8.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 935114
Test Specificity This antibody reacts with ORF-73 of human Herpes virus 8 (HHV8). HHV8 is the likely etiological agent of Kaposi's sarcoma (KS). HHV8 encodes a latent nuclear antigen (LNA) which is the product of the viral gene orf 73. LNA is capable of forming a complex with retinoblastoma susceptibility gene product which may be related to its oncogenic activity.
Target Species Virus
Content And Storage Store at 4C. Do not freeze.
Form Supernatant
Isotype IgG2c
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.