Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Organic Carbon Standard, 20 ppm C, Ricca Chemical
SDP

Catalog No. 1844205 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
1844205 20 L Cubitainer
R18442001C 1 L Amber Glass Bottle
R18442004C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1844205 Supplier Ricca Chemical Company Supplier No. 1844.205
Add to Cart
Add to Cart

Chemical Identifiers

CAS 877-24-7 , 7664-38-2 , 7732-18-5
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Packaging Cubitainer
Concentration or Composition Notes 20ppm C
CAS Min % 98.0, 0.46, 0.0041
CAS Max % 100.0, 0.48, 0.0043
Equivalence 1 mL = 0.020 mg C
Physical Form Liquid
Color Colorless
pH < 2
Boiling Point 100°C
Quantity 20 L
Chemical Name or Material Organic Carbon Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.