Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORL1/OPRL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | ORL1/OPRL1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16914320
![]() |
Novus Biologicals
NBP16914320UL |
20 μL |
Each for $158.00
|
|
|||||
NBP169143
![]() |
Novus Biologicals
NBP169143 |
100 μL |
Each for $487.50
|
|
|||||
Description
ORL1/OPRL1 Polyclonal specifically detects ORL1/OPRL1 in Human samples. It is validated for Western Blot.Specifications
ORL1/OPRL1 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
Kappa-type 3 opioid receptor, KOR-3MGC34578, nociceptin receptor, NOCIR, OORORL1kappa3-related opioid receptor, opiate receptor-like 1, Orphanin FQ receptor | |
OPRL1 | |
IgG | |
41 kDa |
Western Blot | |
Unconjugated | |
RUO | |
P79292 | |
4987 | |
Synthetic peptides corresponding to OPRL1 (opiate receptor-like 1) The peptide sequence was selected from the middle region of OPRL1. Peptide sequence ISVCYSLMIRRLRGVRLLSGSREKDRNLRRITRLVLVVVAVFVGCWTPVQ. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title