Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORMDL1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176577 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176577 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176577 Supplier Novus Biologicals Supplier No. NBP176577
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ORMDL1 Polyclonal specifically detects ORMDL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ORMDL1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_057551
Gene Alias adoplin-1, DKFZp686G141, FLJ44364, ORM1 (S. cerevisiae)-like 1, ORM1-like 1 (S. cerevisiae), ORM1-like protein 1
Gene Symbols ORMDL1
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide of human ORMDL1. The immunogen is located within amino acids 50 - 100 of ORMDL1. Amino Acid Squence: VKGTPFETPDQGKARL
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94101
Test Specificity ORMDL1 antibody may cross-react with other ORMDL protein family members.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.