Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ornithine Carbamoyltransferase Antibody (CL4045), Novus Biologicals™
SDP

Catalog No. NB403060 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB403060 25 μL
NBP259038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB403060 Supplier Novus Biologicals Supplier No. NBP25903825UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ornithine Carbamoyltransferase Monoclonal antibody specifically detects Ornithine Carbamoyltransferase in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen Ornithine Carbamoyltransferase
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:10000 - 1:20000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:10000 - 1:20000
Formulation PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide
Gene Alias EC 2.1.3, EC 2.1.3.3, MGC129967, MGC129968, MGC138856, OCTD, ornithine carbamoyltransferase, ornithine carbamoyltransferase, mitochondrial, Ornithine transcarbamylase, OTCase
Gene Symbols OTC
Host Species Mouse
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ILADYLTLQEHYSSLKGLTLSWIGDGNNILHSIMMSAAKFGMHLQAATPKGYEPDASVTKLAEQYAKENGTKLLLTNDPLEAAHGGNVLITDTWISMGQEEEKKKRLQAFQGYQVTMKTAKVAASDWT
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5009
Test Specificity Specificity of human Ornithine Carbamoyltransferase antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Cynomolgus Monkey
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.