Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ornithine Decarboxylase Antibody (ODC1/2878R), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB384442 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384442 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384442 Supplier Novus Biologicals Supplier No. NBP308791IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Ornithine Decarboxylase Monoclonal antibody specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen Ornithine Decarboxylase
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ODC1/2878R
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1
Host Species Rabbit
Immunogen Recombinant full-length human Ornithine Decarboxylase protein (Uniprot: P11926)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4953
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.