Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orosomucoid 2 Antibody (959445), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB295485 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295485 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295485 Supplier Novus Biologicals Supplier No. FAB9306N

Mouse Monoclonal Antibody

Orosomucoid 2 Monoclonal antibody specifically detects Orosomucoid 2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Orosomucoid 2
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 959445
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AGP 2, AGP2, AGP2alpha-1-acid glycoprotein 2, AGP-B, AGP-B', alpha-1-acid glycoprotein, type 2, OMD 2, orosomucoid 2, orosomucoid-2
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ORM-2, Met1-Glu201, Accession # P19652
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5005
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.