Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORP150/HSP12A Antibody (201), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB096916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096916 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB096916 Supplier Novus Biologicals Supplier No. NBP289890AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ORP150/HSP12A Monoclonal specifically detects ORP150/HSP12A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ORP150/HSP12A
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 201
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias 150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ORP150/HSP12A (NP_001124463.1; Met 695-Leu 994).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 10525
Test Specificity No cross-reactivity in ELISA with: Human Fetuin A
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.