Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORP150/HSP12A Antibody (6E3-2C3), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB259345405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259345405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259345405 Supplier Novus Biologicals Supplier No. NBP259345AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ORP150/HSP12A Monoclonal specifically detects ORP150/HSP12A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ORP150/HSP12A
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 6E3-2C3
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD)
Gene Symbols HYOU1
Host Species Mouse
Immunogen Recombinant Full length GRP170 Protein
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 10525
Test Specificity Detects 170 kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.