Learn More
QuidelOrtho ID-Micro Typing System™ (MTS) Work Table
SDP
Catalog No. okmts9650
Click to view available options
:
MTS Work Table (24 Card)
This item is not returnable. View return policy
This item is not returnable. View return policy
Requires
ID-MTS Manual GEL Workstation™, ORTHO ProVue™
Suitable for a broad range of blood bank applications. This accessory is used with the QuidelOrtho Workstation (OK6904630).
ID-Micro Typing System™
  • Improved sensitivity and specificity
  • No-wash antiglobulin procedure
  • Standardized procedures
  • Improved turnaround time
  • Enhanced regulatory compliance
Antibody screening and identification, ABO blood grouping and Rh phenotyping, Compatibility Testing, Reverse serum grouping, Antigen typing
Specifications
Type MTS™ Work Table
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.