Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ MicroSlides

Click to view available options
:
VITROS 250 Dry Slides: 18-Theophylline
Specifications
Specifications
Sample Type | Heparin, Plasma, Serum |
Type | 18-Theophylline Dry Slide |
Formulation | Alkaline Phosphatase (beef liver, E.C.3.1.3.1) 0.02U, Magnesium Chloride 3μg and p-Nitrophenyl Phosphate 0.3mg, Pigment, Binders, Surfactants, Buffer, Enzyme cofactor, protein, Stabilizer and cross-linking agent |
For Use With (Application) | In-vitro diagnostic use |
Quantity | 5 x 18 Pack |
Sample Size | 10 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction