Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ MicroSlides

Click to view available options
:
VITROS 250 Dry Slides: Potassium
Specifications
Specifications
Sample Type | Heparin, Plasma, Serum, Urine |
Type | Potassium Dry Slide |
Formulation | Silver 0.4mg, Silver Chloride 0.2mg, Sodium Chloride 0.2mg, Potassium Chloride 63μg and Valinomycin 55μg, Binders, Plasticizers, Stabilizer, Surfactants and Nickel-chromium |
For Use With (Application) | In-vitro diagnostic use |
Quantity | 5 x 50 Pack |
Sample Size | 10 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction