Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ MicroSlides

Catalog No. p-3243843
Click to view available options
Quantity:
5 x 18 Pack
5 x 50 Pack
5 x 60 Pack
5/Pk., 90 Slides
Formulation:
1,2-bis(o-Aminophenoxy)ethane-N,N,N',N'-Tetraacetic Acid (Calcium chelator) 242μg, 1,5-bis(2-Hydroxy-3,5-Dichlorophenyl)-3-Cyanoformazan (dye) 38μg, Pigment, Binders, Buffer, dye solubilizer, Surfactants, cross-linking
Alkaline Phosphatase (beef liver, E.C.3.1.3.1) 0.02U, Magnesium Chloride 3μg and p-Nitrophenyl Phosphate 0.3mg, Pigment, Binders, Surfactants, Buffer, Enzyme cofactor, protein, Stabilizer and cross-linking agent
Copper Sulfate 0.9mg, Tartaric Acid 1.2mg and Lithium Hydroxide 1.3mg, Polymer beads, Binders and Surfactants
Not Applicable
Silver 0.4mg, Silver Chloride 0.2mg, Sodium Chloride 0.2mg, Potassium Chloride 63μg and Valinomycin 55μg, Binders, Plasticizers, Stabilizer, Surfactants and Nickel-chromium
Silver 0.4mg, Silver Chloride 0.2mg, Sodium Chloride 0.3mg, Methyl Monensin 50μg, Binders, Buffer, Plasticizers, Stabilizer, Surfactants and Nickel-chromium
Uricase (Candida utilis, E.C.1.7.3.3) 0.02U, Peroxidase (horseradish root, E.C.1.11.1.7) 0.6U and 2-(3,5-Dimethoxy-4-Hydroxyphenyl)-4,5-bis-(4-Dimethylaminophenyl) Imidazole (leucodye) 14μg, Stabilizers, Pigment, Binders, Buffer, Surfact
p-Methylaminophenol Sulfate 350μg, Ammonium Molybdate 340μg, Pigment, Binders, Surfactants, Buffer and Stabilizers
Type:
18-Magnesium Dry Slide
18-Theophylline Dry Slide
Direct HDL Cholesterol-dHDL
Magnesium Dry Slide
Phosphorus Dry Slide
Potassium Dry Slide
Sodium Dry Slide
Total Protein Dry Slide
Uric Acid Dry Slide

For use with the QuidelOrtho VITROS™ 250 System

Specifications

Sample Type Heparin, Plasma, Serum, Urine
Type 18-Magnesium Dry Slide
Formulation 1,2-bis(o-Aminophenoxy)ethane-N,N,N',N'-Tetraacetic Acid (Calcium chelator) 242μg, 1,5-bis(2-Hydroxy-3,5-Dichlorophenyl)-3-Cyanoformazan (dye) 38μg, Pigment, Binders, Buffer, dye solubilizer, Surfactants, cross-linking
For Use With (Application) In-vitro diagnostic use
Quantity 5 x 18 Pack
Sample Size 5 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.