Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ortho-Phosphoric acid 85%, Suprapur™, MilliporeSigma™
GSA_VA

Catalog No. p-4640352 Shop All MilliporeSigma Products
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L

Catalog No. M1005520250

Supplier: MilliporeSigma 1.00552.0250

Only null left
Add to Cart
Add to Cart

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid

Specifications

Color Colorless
Density 1.71g/cm3 (20°C)
Packaging Plastic Bottle
Vapor Pressure 2 hPa (20°C)
Assay Percent Range 85%
Quantity 250 mL
Physical Form Liquid
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
Solubility Information Soluble (2°C)
SMILES OP(=O)(O)O
Molecular Weight (g/mol) 97.994
Grade Suprapur
Chemical Name or Material Suprapur™
Show More Show Less
Hazard Category SignalWord: Danger
Recommended Storage Store above +15°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.