Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ortho-Phosphoric Acid, MilliporeSigma

Catalog No. M1005730510 Shop All MilliporeSigma Products
Click to view available options
Quantity:
0.5 L
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

85%, for analysis EMSURE™ ACS,ISO,Reag. Ph Eur

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Weight (g/mol) 98 g/mol

Specifications

Packaging Plastic bottle
Quantity 0.5 L
Percent Purity 85%
Grade ACS
Chemical Name or Material Ortho-Phosphoric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.