Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OS9 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB100519G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100519G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100519G Supplier Novus Biologicals Supplier No. NB100519G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OS9 Polyclonal specifically detects OS9 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen OS9
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q13438
Gene Alias Amplified in osteosarcoma 9, endoplasmic reticulum lectin 2, ERLEC2, erlectin 2, OS-9, osteosarcoma amplified 9, endoplasmic reticulum associated protein, osteosarcoma amplified 9, endoplasmic reticulum lectin, protein OS-9
Gene Symbols OS9
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of isoform 1 of the human OS9 protein (within residues 300-400). [Swiss-Prot# Q13438]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Hypoxia, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10956
Test Specificity This is specific for isoforms 1 and 2 of human OS-9.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.