Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OS9 Antibody, Novus Biologicals™
SDP

Catalog No. BC100520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BC100520 0.1 mL
BC100520SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BC100520 Supplier Novus Biologicals Supplier No. BC100520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

OS9 Polyclonal specifically detects OS9 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, KnockDown, Simple Western, Western Blot.

Specifications

Antigen OS9
Applications Immunocytochemistry, Immunofluorescence, KnockDown, Simple Western, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:100, Immunocytochemistry/Immunofluorescence 1:500
Formulation Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
Gene Accession No. Q13438
Gene Alias Amplified in osteosarcoma 9, endoplasmic reticulum lectin 2, ERLEC2, erlectin 2, OS-9, osteosarcoma amplified 9, endoplasmic reticulum associated protein, osteosarcoma amplified 9, endoplasmic reticulum lectin, protein OS-9
Gene Symbols OS9
Host Species Rabbit
Immunogen A C-terminal synthetic peptide made to isoform 1 of the human OS9 protein sequence (between residues 600-667). There are 3 isoforms of this protein. The immunogen used for this antibody is conserved in isoforms 1, 2, and 3 of the human OS9 protein.
Molecular Weight of Antigen 75 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Hypoxia, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10956
Test Specificity Human, mouse and rat (see customer review).
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.