Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSBP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001500 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001500 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001500 Supplier Novus Biologicals Supplier No. NB1001500
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 5 publications

OSBP1 Polyclonal specifically detects OSBP1 in Human samples. It is validated for Immunoprecipitation, Peptide ELISA, Western Blot.

Specifications

Antigen OSBP1
Applications Immunoprecipitation, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Immunoprecipitation, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P22059
Gene Alias OSBP1oxysterol-binding protein 1, oxysterol binding protein
Gene Symbols OSBP
Host Species Goat
Immunogen Peptide with sequence C-KHQLEETKKEKRTRIP corresponding to internal region according to NP_002547.1.
Molecular Weight of Antigen 89 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5007
Test Specificity Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.