Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSBPL10 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002448 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002448 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002448 Supplier Novus Biologicals Supplier No. NB1002448
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

OSBPL10 Polyclonal specifically detects OSBPL10 in Human, Mouse samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen OSBPL10
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_060254.2
Gene Alias ORP-10, ORP10FLJ20363, OSBP9, OSBP-related protein 10, oxysterol binding protein-like 10, oxysterol-binding protein-related protein 10
Gene Symbols OSBPL10
Host Species Goat
Immunogen Peptide with sequence C-DAATEQKRHLEEKQ corresponding to internal region according to NP_060254.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 114884
Test Specificity Cross reacts with Human and Mouse. Expected from sequence similarity: Rat, Dog, Pig and Cow.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.