Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSBPL11 Antibody (OTI6A6), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP273160A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273160A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273160A30 Supplier Novus Biologicals Supplier No. NBP273160AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OSBPL11 Monoclonal specifically detects OSBPL11 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen OSBPL11
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI6A6
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ13012, FLJ13164, ORP11OSBP-related protein 11, ORP-11TCCCIA00292, OSBP12, oxysterol binding protein-like 11, oxysterol-binding protein-related protein 11
Gene Symbols OSBPL11
Host Species Mouse
Immunogen Full length recombinant protein of human OSBPL11 (NP_073613) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 114885
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.