Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSBPL2 Antibody (OTI2E6), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N273161A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273161A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273161A647 Supplier Novus Biologicals Supplier No. NBP273161AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OSBPL2 Monoclonal specifically detects OSBPL2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen OSBPL2
Applications Western Blot
Classification Monoclonal
Clone OTI2E6
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias KIAA0772FLJ20223, ORP-2MGC4307, ORP2MGC8342, OSBP-related protein 2, oxysterol binding protein-like 2, oxysterol-binding protein-like 2, oxysterol-binding protein-related protein 2
Gene Symbols OSBPL2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 258-480 of human OSBPL2 (NP_653081) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9885.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.