Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSBPL7 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001335 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001335 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001335 Supplier Novus Biologicals Supplier No. NB1001335
Add to Cart
Add to Cart

Goat Polyclonal Antibody

OSBPL7 Polyclonal specifically detects OSBPL7 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA.

Specifications

Antigen OSBPL7
Applications Immunocytochemistry, Immunofluorescence, Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q9BZF2
Gene Alias ORP-7, ORP7MGC71150, OSBP-related protein 7, oxysterol binding protein-like 7, oxysterol-binding protein-related protein 7
Gene Symbols OSBPL7
Host Species Goat
Immunogen Peptide with sequence C-EPGYGNMDGAVLW corresponding to C-Terminus according to NP_665741.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 114881
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.