Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSCAR Antibody (MM0502-6S6), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP211835IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211835IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211835IR Supplier Novus Biologicals Supplier No. NBP211835IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OSCAR Monoclonal specifically detects OSCAR in Human samples. It is validated for Western Blot.

Specifications

Antigen OSCAR
Applications Western Blot
Classification Monoclonal
Clone MM0502-6S6
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias hOSCAR, MGC33613, osteoclast associated receptor OSCAR-S1, osteoclast associated receptor OSCAR-S2, osteoclast associated, immunoglobulin-like receptor, osteoclast-associated immunoglobulin-like receptor, Osteoclast-associated receptor, PIgR-3, PIGR3, Poly-Ig receptor 3, Polymeric immunoglobulin receptor 3
Gene Symbols OSCAR
Host Species Mouse
Immunogen Human recombinant OSCAR (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 126014
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.