Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSCAR Monoclonal Antibody (C2), Invitrogen™
Mouse Monoclonal Antibody
$243.00 - $522.50
Specifications
Antigen | OSCAR |
---|---|
Clone | C2 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543635
|
Invitrogen™
MA543635 |
20 μL |
Each for $243.00
|
|
|||||
PIMA543634
|
Invitrogen™
MA543634 |
100 μL |
Each for $522.50
|
|
|||||
Description
Recently, a novel member of the leukocyte receptor complex (LRC) encoded family has been identified that is expressed specifically in osteoclast (OC). This protein names OC-associated receptor or OSCAR, play critical roles in the regulation of both innate and adaptive immune responses and bone-specific regulator of OC differentiation. Osteoclasts are multinucleated cells that resorb bone and are essential for bone homeostasis. This protein family plays critical roles in the regulation of both innate and adaptive immune responses. Different from the other LRC members, OSCAR expression is detected specifically in preosteoclasts or mature osteoclasts. OSCAR may be an important bone-specific regulator of osteoclast differentiation. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
OSCAR | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
MGC33613; mOSCAR; mOSCAR-M1; mOSCAR-M2; mOSCAR-M3; ocl178; Oscar; osteoclast associated receptor; osteoclast associated receptor OSCAR-S1; osteoclast associated receptor OSCAR-S2; osteoclast associated, immunoglobulin-like receptor; osteoclast-associated immunoglobulin-like receptor; Osteoclast-associated receptor; osteoclast-associated receptor OSCAR; PIGR3; PIgR-3; poly-Ig receptor 3; polymeric immunoglobulin receptor 3 | |
OSCAR | |
IgG1 κ | |
Protein A/G | |
Antibody |
C2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q8IYS5 | |
126014 | |
Polypeptide conjugated human OVA. The polypeptide sequence is QGGIYRCCYRRPDWGP. | |
Primary | |
-20°C | |
OSCAR |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title