Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSER1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$480.74

Specifications

Antigen C20orf111
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP156864
SDP
View Documents
Novus Biologicals
NBP156864
100 μL
Each for $480.74
Only null left
Add to Cart
 
NBP15686420
SDP
View Documents
Novus Biologicals
NBP15686420UL
20 μL N/A N/A N/A
Description

Description

OSER1 Polyclonal specifically detects OSER1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

C20orf111
Polyclonal
Rabbit
Q9NX31
51526
Synthetic peptides corresponding to C20ORF111 The peptide sequence was selected from the N terminal of C20ORF111. Peptide sequence RGAVRTQRRRRSKSPVLHPPKFIHCSTIASSSSSQLKHKSQTDSPDGSSG.
Primary
Western Blot
Unconjugated
RUO
chromosome 20 open reading frame 111, dJ1183I21.1, HSPC207, Osr1, oxidative stress responsive 1, Perit1, peroxide-inducible transcript 1
C20ORF111
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.