Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSGEP Antibody (OTI9E3), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N273163APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273163APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273163APC7 Supplier Novus Biologicals Supplier No. NBP273163APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OSGEP Monoclonal specifically detects OSGEP in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.

Specifications

Antigen OSGEP
Applications Flow Cytometry
Classification Monoclonal
Clone OTI9E3
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.4.24.57, GCPL1FLJ20411, hOSGEP, KAE1, OSGEP1, O-sialoglycoprotein endopeptidase, probable O-sialoglycoprotein endopeptidase, PRSMG1, t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEP
Gene Symbols OSGEP
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-335 of human OSGEP (NP_060277) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55644.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.