Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSMR beta Antibody (118125) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. NB376107 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376107 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376107 Supplier Novus Biologicals Supplier No. FAB662J
Add to Cart
Add to Cart

Rat Monoclonal Antibody

OSMR beta Monoclonal antibody specifically detects OSMR beta in Mouse samples. It is validated for Western Blot, Immunocytochemistry

Specifications

Antigen OSMR beta
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 118125
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse OSM R beta , Glu24-Leu738, Accession # O70458
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9180
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.