Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSMR beta Antibody (469221), Biotin, Novus Biologicals™
SDP

Catalog No. NB370234 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370234 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370234 Supplier Novus Biologicals Supplier No. FAB4389B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OSMR beta Monoclonal specifically detects OSMR beta in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry, Gene Knock-Out, Western Blot.

Specifications

Antigen OSMR beta
Applications CyTOF, ELISA, Flow Cytometry, Gene Knock-Out, Western Blot
Classification Monoclonal
Clone 469221
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, ELISA, CyTOF-ready, Knockout Validated
Formulation PBS
Gene Alias IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human OSM R beta , Glu28-Ser739, Accession # Q99650
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9180
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.