Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSMR beta Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$158.00 - $487.50

Specifications

Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16254320
SDP
View Documents
Novus Biologicals
NBP16254320UL
20 μL
Each for $158.00
Only null left
Add to Cart
 
NBP162543
SDP
View Documents
Novus Biologicals
NBP162543
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

OSMR beta Polyclonal specifically detects OSMR beta in Human samples. It is validated for Western Blot.
Specifications

Specifications

Western Blot
Unconjugated
RUO
IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMRBMGC150626
OSMR
IgG
This product is specific to Subunit or Isoform: beta.
Polyclonal
Rabbit
Q99650
9180
Synthetic peptides corresponding to OSMR(oncostatin M receptor) The peptide sequence was selected from the N terminal of OSMR. Peptide sequence YQSEVLAERLPLTPVSLKVSTNSTRQSLHLQWTVHNLPYHQELKMVFQIQ.
Primary
110 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.