Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OspA Antibody, Novus Biologicals™
SDP

Catalog No. NBP178112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178112 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178112 Supplier Novus Biologicals Supplier No. NBP178112
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OspA Polyclonal specifically detects OspA in Bacteria samples. It is validated for Western Blot, ELISA.

Specifications

Antigen OspA
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:5000
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P0CL66
Gene Alias outer surface protein A
Gene Symbols IL12A
Host Species Rabbit
Immunogen MBP-fusion protein corresponding to Borrelia burgdorferi OspA protein. (Uniprot: P0CL66)
Molecular Weight of Antigen 29.37 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3592
Test Specificity This antibody is specific for Borrelia burgdorferi OspA protein. A BLAST analysis was used to suggest cross-reactivity with OspA from B. burgdorferi and sources based on 100% identity with the immunizing sequence.
Reconstitution Reconstitute with 100 ul deionized water (or equivalent)
Target Species Bacteria
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.